Skip to Content
Merck
All Photos(1)

Documents

880400P

Avanti

16:0 PEG350 PE

Avanti Research - A Croda Brand 880400P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-350] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 200 mg (880400P-200mg)
pkg of 1 × 25 mg (880400P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880400P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

General description

16:0 PEG350 PE is a polyethylene glycol (PEG) conjugated phospholipid. It is a biopolymer with a molecular weight of 1075.4 Da.

Application

16:0 PEG350 PE is suitable for use in the preparation of Pdots and to differentiate based on shell′s composition. It might also be used in membrane preparation to study water dynamics.

Biochem/physiol Actions

Polyethylene glycol (PEG) conjugated phospholipids are useful in liposome formulation for maintaining their stereochemistry. They also serve as drug carriers. PEGylated lipids aid in the formation of mixed micelles at high concentration.

Packaging

20 mL Clear Glass Screw Cap Vial (880400P-200mg)
5 mL Clear Glass Sealed Ampule (880400P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo M Clop et al.
The journal of physical chemistry. B, 118(23), 6150-6158 (2014-05-09)
The present work was aimed at studying the molecular dynamics at different levels of model membranes having a simulated glycoclix, with focus on the molecular crowding conditions at the lipid-water interfacial region. Thus, binary mixtures of dipalmitoylphosphatidylcholine (dpPC) and a
Pdots nanoparticles load photosensitizers and enhance efficiently their photodynamic effect by FRET
Haimov E, et al.
Royal Society of Chemistry Advances, 5(24), 18482-18491 (2015)
Sirkku Hanski et al.
Biomacromolecules, 11(12), 3440-3447 (2010-10-27)
We report on highly ordered oblique self-assemblies in ionic complexes of PEGylated triple-tail lipids and cationic polypeptides, as directed by side-chain crystallization, demonstrating also reversible oblique-to-hexagonal order-order transitions upon melting of the side chains. This is achieved in bulk by
G F Peacock et al.
Drug delivery, 10(1), 29-34 (2003-01-30)
A new cholesterol-carborane conjugate (BCH) has been synthesized as a potential targeting agent for boron neutron capture therapy (BNCT) of cancers. The compound is extremely water insoluble and was formulated in two liposomal formulations to determine if the compound could
Maria C Sandström et al.
Langmuir : the ACS journal of surfaces and colloids, 23(8), 4192-4198 (2007-03-09)
Polyethylene glycol (PEG)-conjugated lipids are commonly employed for steric stabilization of liposomes. When added in high concentrations PEG-lipids induce formation of mixed micelles, and depending on the lipid composition of the sample, these may adapt either a discoidal or a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service