Skip to Content
Merck
All Photos(1)

Key Documents

860637P

Avanti

Galactosyl(β) Sphingosine-d5

Avanti Research - A Croda Brand 860637P, powder

Synonym(s):

D-galactosyl-β1-1′-D-erythro-sphingosine-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42D5NO7
CAS Number:
Molecular Weight:
466.66
UNSPSC Code:
12352211
NACRES:
NA.12

form

powder

packaging

pkg of 1 × 1 mg (860637P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860637P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCC([2H])(C([2H])([2H])[2H])[2H])(O)[C@](N)([H])CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI key

HHJTWTPUPVQKNA-LTNFHSIGSA-N

Application

Galactosyl(β) Sphingosine-d5 has been used as an internal standard for the quantification of galactosylsphingosine, in the extracts from the mouse cerebellum by liquid chromatography-tandem mass spectrometry.

Biochem/physiol Actions

Galactosylsphingosine, also known as psychosine, has an ability to induce apoptosis in various types of cells including oligodendrocytes. It is a toxic metabolite. Accumulation of this cytotoxic sphingolipid in the brain due to the deficiency of galactocerebrosidase (GALC) leads to the development of Krabbe disease.

Packaging

5 mL Amber Glass Screw Cap Vial (860637P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacqueline A Hawkins-Salsbury et al.
Journal of lipid research, 54(12), 3303-3311 (2013-09-06)
Globoid cell leukodystrophy (GLD) is a neurological disease caused by deficiency of the lysosomal enzyme galactosylceramidase (GALC). In the absence of GALC, the cytotoxic glycosphingolipid, psychosine (psy), accumulates in the nervous system. Psychosine accumulation preferentially affects oligodendrocytes, leading to progressive
Victoria Schiffer et al.
PloS one, 15(1), e0227077-e0227077 (2020-01-14)
Gaucher disease is caused by a deficiency in glucocerebrosidase that can result in non-neuronal as well as neuronal symptoms. Common visceral symptoms are an increased organ size, specifically of the spleen, and glucosylceramide as well as glucosylsphingosine substrate accumulations as
Catherine O'Sullivan et al.
Journal of cell science, 128(21), 3878-3887 (2015-09-12)
Globoid cell leukodystrophy (Krabbe disease) is a rare infantile neurodegenerative disorder. Krabbe disease is caused by deficiency in the lysosomal enzyme galactocerebrosidase (GALC) resulting in accumulation, in the micromolar range, of the toxic metabolite galactosylsphingosine (psychosine) in the brain. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service