Skip to Content
Merck
All Photos(1)

Key Documents

860628P

Avanti

C24 Dihydroceramide (d18:0/24:0)

Avanti Research - A Croda Brand 860628P, powder

Synonym(s):

N-lignoceroyl-D-erythro-sphinganine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H85NO3
CAS Number:
Molecular Weight:
652.13
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 10 mg (860628P-10mg)
pkg of 1 × 25 mg (860628P-25mg)
pkg of 1 × 5 mg (860628P-5mg)
pkg of 1 × 50 mg (860628P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860628P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCCCCCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI

1S/C42H85NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-42(46)43-40(39-44)41(45)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h40-41,44-45H,3-39H2,1-2H3,(H,43,46)/t40-,41+/m0/s1

InChI key

BPLYVSYSBPLDOA-WVILEFPPSA-N

Biochem/physiol Actions

Dihydroceramide acts as a precursor for ceramide synthesis. It has an ability to induce autophagy in the perinuclear region. Dihydroceramide affects autophagic flux and enhances the formation of lipid droplets.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860628P-10mg)
5 mL Amber Glass Screw Cap Vial (860628P-25mg)
5 mL Amber Glass Screw Cap Vial (860628P-50mg)
5 mL Amber Glass Screw Cap Vial (860628P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dihydroceramide is a key metabolite that regulates autophagy and promotes fibrosis in hepatic steatosis model
Lee Ah young, et al.
Biochemical and biophysical research communications, 494(3-4), 460-469 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service