Skip to Content
Merck
All Photos(1)

Key Documents

856706P

Avanti

13:0 Lyso PE

1-tridecanoyl-sn-glycero-3-phosphoethanolamine, powder

Synonym(s):

1-tridecanoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine; PE(13:0/0:0); 110696

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H38NO7P
CAS Number:
Molecular Weight:
411.47
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (may contain up to 10% of the 2-LPE isomer, TLC)

form

powder

packaging

pkg of 1 × 5 mg (856706P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 856706P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[NH3+])=O)([H])COC(CCCCCCCCCCCC)=O

InChI key

IJTWJFQIBBDJJI-QGZVFWFLSA-N

Application

13:0 Lyso PE has been used as an internal standard for the quantification of glycerophospholipids.

Packaging

5 mL Amber Glass Screw Cap Vial (856706P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversed phase LC/MS/MS method for targeted quantification of glycerophospholipid molecular species in plasma
Uhl O, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 879(30), 3556-3564 (2011)
Untargeted lipidomic analysis of plasma from high fat diet-induced obese rats using UHPLC- linear trap quadrupole -Orbitrap MS.
Siddabasave Gowda B Gowda et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2020-01-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service