Skip to Content
Merck
All Photos(2)

Key Documents

850705P

Avanti

16:0 PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine; DPPE; PE(16:0/16:0); 110633

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H74NO8P
CAS Number:
Molecular Weight:
691.96
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850705P-1g)
pkg of 1 × 200 mg (850705P-200mg)
pkg of 1 × 25 mg (850705P-25mg)
pkg of 1 × 500 mg (850705P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C37H74NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(34-45-47(41,42)44-32-31-38)46-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35H,3-34,38H2,1-2H3,(H,41,42)/t35-/m1/s1

InChI key

SLKDGVPOSSLUAI-PGUFJCEWSA-N

General description

Phosphatidylethanolamine is an aminophospholipid. It is present in eukaryotic and prokaryotic cells. Phosphatidylethanolamine is one of the most abundant glycerophospholipids in biological membranes. It is found in nervous tissue.

Application

16:0 PE has been used:
  • for the preparation of perfluorocarbon (PFC) nanoparticles
  • for lipid binding by liposome flotation assays
  • to characterize the metabolic fate of synaptamide in a dopaminergic cell line

Biochem/physiol Actions

Phosphatidylethanolamine forms pure lipid structures. It plays a vital role in autophagy, cell division and protein folding. Phosphatidylethanolamine functions as a precursor for the synthesis of various protein modifications. It controls membrane fluidity in eukaryotic cells. Phosphatidylethanolamine behaves as an intermediate for the synthesis of glycerophospholipid classes.

Packaging

20 mL Clear Glass Screw Cap Vial (850705P-1g)
20 mL Clear Glass Screw Cap Vial (850705P-500mg)
5 mL Amber Glass Screw Cap Vial (850705P-200mg)
5 mL Amber Glass Screw Cap Vial (850705P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Introduction to Biological Membranes: Composition, Structure and Function (2016)
The Membranes of Cells (2016)
Anti-JNK2 peptide-siRNA nanostructures improve plaque endothelium and reduce thrombotic risk in atherosclerotic mice
Pan H, et al.
International journal of nanomedicine, 13, 5187-5187 (2018)
Metabolic studies of synaptamide in an immortalized dopaminergic cell line
Sonti S, et al.
Prostaglandins & other lipid mediators, 141, 25-33 (2019)
Shilpa Sonti et al.
Prostaglandins & other lipid mediators, 141, 25-33 (2019-02-15)
Synaptamide, the N-acylethanolamine of docosahexaenoic acid (DHA), is structurally similar to the endocannabinoid N-arachidonoylethanolamine, anandamide. It is an endogenous ligand at the orphan G-protein coupled receptor 110 (GPR110; ADGRF1), and induces neuritogenesis and synaptogenesis in hippocampal and cortical neurons, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service