Skip to Content
Merck
All Photos(3)

Documents

841199P

Avanti

E. coli CA

Avanti Research - A Croda Brand 841199P, powder

Synonym(s):

Cardiolipin (E. coli) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (841199P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 841199P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OC[C@]([H])(OC(CCCCCCCC1C(C1)CCCCCCC)=O)COC(CCCCCCCCCCCCCC)=O)OCC([H])(O)COP([O-])(OC[C@]([H])(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCC/C=C/CCCCCCCC)=O)=O.[Na+].[Na+]

General description

Cardiolipin is the most important phospholipid of mitochondria. E.coli membrane consists 5-10% of cardiolipin. Cardiolipin is a non-bilayer anionic phospholipid, which has four acyl chains and small headgroup.

Application

E. coli CA has been used for western blotting. It has also been used for lipid monolayer assay.

Biochem/physiol Actions

Cardiolipin plays a major role in the dynamic organization of membranes and in cell division. It is implicated in proton uptake pathway.

Packaging

5 mL Clear Glass Sealed Ampule (841199P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrobiologia, 104(3), 472-486 (2018)
Exclusion of assembled M re B by anionic phospholipids at cell poles confers cell polarity for bidirectional growth
Kawazura T, et al.
Molecular Microbiology, 104(3), 472-486 (2017)
Javier Hoyo et al.
General physiology and biophysics, 39(2), 195-202 (2020-04-25)
The current strategies to eradicate bacteria require that the antimicrobial agent either penetrate or disrupt the bacterial membrane. In Escherichia coli (E.coli) as a model of Gram-negative strains, the antimicrobials have to cross two barriers - the outer and the
Mikhail Bogdanov et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(34), 15057-15062 (2010-08-11)
Phosphatidylcholine (PC) has been widely used in place of naturally occurring phosphatidylethanolamine (PE) in reconstitution of bacterial membrane proteins. However, PC does not support native structure or function for several reconstituted transport proteins. Lactose permease (LacY) of Escherichia coli, when
Functional role of cardiolipin in mitochondrial bioenergetics
Paradies G, et al.
Biochimica et Biophysica Acta - Bioenergetics, 1837(4), 408-417 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service