Skip to Content
Merck
All Photos(2)

Key Documents

840420P

Avanti

18:1 Phosphatidylmethanol

1,2-dioleoyl-sn-glycero-3-phosphomethanol (sodium salt), powder

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphomethanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H74O8PNa
CAS Number:
Molecular Weight:
736.97
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840420P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840420P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCC\C=C/CCCCCCCC)OC

InChI

1S/C40H75O8P.Na/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-39(41)46-36-38(37-47-49(43,44)45-3)48-40(42)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2;/h18-21,38H,4-17,22-37H2,1-3H3,(H,43,44);/q;+1/p-1/b20-18-,21-19-;/t38-;/m1./s1

InChI key

VZVXMZAFOPVENO-MDTXTNHHSA-M

General description

18:1 Phosphatidylmethanol (1,2-dioleoyl-sn-glycero-3-phosphomethanol) is a phospholipid which carries an anionic head group. It is categorized as an alkyl phosphate under lipd headgroup modified lipids.

Application

18:1 Phosphatidylmethanol (1,2-dioleoyl-sn-glycero-3-phosphomethanol) may be used in unilamellar vesicles for fluorescence quenching vesicle binding assay and as an internal standard in ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) analysis for the quantification of phosphatidylethanols (PEths) in whole blood, venous/capillary-dried blood spots (V-DBS and C-DBS).

Packaging

5 mL Clear Glass Sealed Ampule (840420P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian Jeppesen et al.
The Journal of biological chemistry, 283(10), 6126-6135 (2008-01-05)
Annexin V binds to membranes with very high affinity, but the factors responsible remain to be quantitatively elucidated. Analysis by isothermal microcalorimetry and calcium titration under conditions of low membrane occupancy showed that there was a strongly positive entropy change
Natalie Kummer et al.
Analytical and bioanalytical chemistry, 408(3), 825-838 (2015-11-26)
Phosphatidylethanol species (PEths) are promising biomarkers of alcohol consumption. Here, we report on the set-up, validation, and application of a novel UHPLC-ESI-MS/MS method for the quantification of PEth 16:0/18:1, PEth 18:1/18:1, and PEth 16:0/16:0 in whole blood (30 μL) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service