Skip to Content
Merck
All Photos(2)

Key Documents

810132C

Avanti

18:1-06:0 NBD PC

Avanti Research - A Croda Brand 810132C

Synonym(s):

1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H64N5O11P
CAS Number:
Molecular Weight:
797.92
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810132C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810132C

concentration

1 mg/mL (810132C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphocholine is considered as a precursor molecule. It is formed during the breakdown of phosphatidylcholine (PC) metabolism. 18:1-06:0 NBD PC is a fluorescent lipid analog/probe.[1][2]

Application

18:1-06:0 NBD PC is suitable for use:
  • as a component in preparing lipid solution for bilayer formation[1]
  • to use as a single donor in fluorescence resonance energy transfer (FRET) based assay[2]
  • to prepare phospholipid-coated beads[3]

Biochem/physiol Actions

NBD‐PC can be used to probe the existence of (Rho‐labeled) protease‐activated receptor (PAR) pepducins in the outer and inner leaflets.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810132C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Large scale biomimetic membrane arrays
Hansen JS, et al.
Analytical and Bioanalytical Chemistry, 395(3), 719-727 (2009)
Coordinated balance of Rac1 and RhoA plays key roles in determining phagocytic appetite
Kim SY, et al.
PLoS ONE, 12(4), e0174603-e0174603 (2017)
Yuta Ryoden et al.
Journal of immunology (Baltimore, Md. : 1950), 204(3), 559-568 (2019-12-22)
In response to extracellular ATP, the purinergic receptor P2X7 mediates various biological processes, including phosphatidylserine (PtdSer) exposure, phospholipid scrambling, dye uptake, ion transport, and IL-1β production. A genome-wide CRISPR screen for molecules responsible for ATP-induced PtdSer exposure identified a transmembrane
Anticoagulant and antithrombotic properties of intracellular protease-activated receptor antagonists
Wielders SJH, et al.
Journal of Thrombosis and Haemostasis, 5(3), 571-576 (2007)
Johan Tornmalm et al.
Scientific reports, 9(1), 18133-18133 (2019-12-04)
Protein-lipid interactions in cellular membranes modulate central cellular functions, are often transient in character, but occur too intermittently to be readily observable. We introduce transient state imaging (TRAST), combining sensitive fluorescence detection of fluorophore markers with monitoring of their dark

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service