Skip to Content
Merck
All Photos(1)

Key Documents

Z743733

ART® Low Retention Barrier Tip

ART 10E, size 100 μL, racked, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121603
NACRES:
NB.02
manufacturer/tradename:
Molecular BioProducts 2065E-05
size:
100 μL
sterility:
sterile
feature:
ART 10E
racked
packaging:
pkg of 5X960 ea

sterility

sterile

feature

ART 10E
racked

packaging

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 2065E-05

technique(s)

PCR: suitable

size

100 μL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

ART® Low Retention Barrier Tips are specially designed sterile pipette tips to eliminate sample retention.

Features and Benefits

  • Extremely hydrophobic inner surface enhances sample reproducibility and accuracy.
  • Crystal clear tips provide excellent visibility for transparent samples.
  • Tips form a secure seal when exposed to potential contaminants.
  • Certified RNase, DNase, ATP, endotoxin, and pyrogen free for reliable and contamination-free performance.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service