Z742362
2MAG FABdrive strong magnetic stirrer for up to 1,000 L
for use with external FABcontrol control unit
Synonym(s):
high volume stirrer, induction plate, induction stirrer, laboratory stirrer, magnetic stir plate, magnetic stirrer
About This Item
Recommended Products
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
2mag 50101
parameter
-10-50 °C temp. limit (at 95% humidity)
80-990 rpm speed
W × D × H
180 mm × 180 mm × 75 mm
plate size × W × L
× 180 mm × 180 mm
stirrer volume
1-1000 mL , per stirring position
weight
9.0 kg
Related Categories
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service