Z677531
Lapren™ latex/chloroprene flock lined gloves
size S
Synonym(s):
chemically resistant glove
About This Item
Recommended Products
material
green (chloroprene, natural latex)
green gloves
size
S
sterility
non-sterile
feature
CE compliant
disposable
latex free: no
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
packaging
package of 100 ea
manufacturer/tradename
KCL 706 - 07
KCL 706
L × thickness
310 mm × 0.6 mm
General description
With cotton flock lining, stretched cuff and anatomical design, this product meets all requirements of comfort and hygiene.
- Ergonomic fit and high elasticity ensures good sensitivity
- Profiled palm for firm grip when wet
- High mechanical resistance
- Cotton flock lining improves comfort
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- CE compliant
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)