Key Documents
Z281999
Coors alumina mortar
O.D. × H 50 mm × 26 mm, capacity 7 mL
About This Item
UNSPSC Code:
41101702
NACRES:
NB.35
material
alumina
manufacturer/tradename
Coorstek 60358
O.D. × H
50 mm × 26 mm
capacity
7 mL
Related Categories
General description
99.5% alumina. Extremely hard and completely nonabsorbent surface. Completely glazed except for inside grinding surface. Order pestle separately.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)