Key Documents
Z151661
Sigma-Aldrich
"T"-purge valve
CGA Inlet size 180
About This Item
UNSPSC Code:
40141600
NACRES:
NB.51
Pricing and availability is not currently available.
pressure measuring range
≤3,000 psig
CGA Inlet size
180
General description
  • Use with SS lecture-bottle regulators when regulating corrosive gases
  • 316 SS with CGA inlet/outlet connections
  • Diaphragm-seal purge valve with 1/4 in. NPTM inlet
  • Check-valve at purge-inlet prevents backflow of gas
Valve installs between lecture-bottle and regulator to purge system of air, moisture, and process gas. Minimizes gas hydrolysis and corrosion damage to equipment.
Purge system with dry inert gas before introducing corrosive gases and again immediately after use. PTFE sealing washers are NOT included.
Note: Order spare washers, listed separately.
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)