Z150096
Pressure relief valve for Kilo-Lab® cylinders
brass, NPTM 1/4 in.
All Photos(1)
About This Item
UNSPSC Code:
40141600
NACRES:
NB.43
Recommended Products
material
brass
NPTM
1/4 in.
General description
Set to release at 200psi.
Legal Information
Kilo-Lab is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)