Skip to Content
Merck
All Photos(1)

Key Documents

W389706

Sigma-Aldrich

2-Propanethiol

≥95%

Synonym(s):

Isopropyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHSH
CAS Number:
Molecular Weight:
76.16
FEMA Number:
3897
Beilstein:
605260
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.197
NACRES:
NA.21
Organoleptic:
meaty; onion; sulfurous
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

vapor density

2.6 (vs air)

vapor pressure

455 mmHg ( 37.8 °C)

Assay

≥95%

refractive index

n20/D 1.426 (lit.)

bp

57-60 °C (lit.)

mp

−131 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; onion; sulfurous

SMILES string

CC(C)S

InChI

1S/C3H8S/c1-3(2)4/h3-4H,1-2H3

InChI key

KJRCEJOSASVSRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Propanethiol is a volatile sulfur compound that is reported to occur in irradiated beef.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-23.8 °F - closed cup

Flash Point(C)

-31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irradiation flavor and the volatile components of beef.
Wick EL, et al.
Advances in Chemistry Series, 12?25, 12?25-12?25 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service