W36691
D-Glucose-13C6
API for Clinical Studies, 99 atom % 13C
Synonym(s):
D-Glucose-ul-13C, Dextrose-13C6
About This Item
Recommended Products
grade
cGMP
isotopic purity
99 atom % 13C
Assay
99% (CP)
optical activity
[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)
mp
150-152 °C (lit.)
mass shift
M+6
storage temp.
15-25°C
SMILES string
O[13CH2][13C@H]1O[13C@H](O)[13C@H](O)[13C@@H](O)[13C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1
InChI key
WQZGKKKJIJFFOK-NYKRWLDMSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service