W248606
Fenugreek absolute
natural, FG
Synonym(s):
Trigonella foenum-graecum oleoresin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
FEMA Number:
2486
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
Halal
Kosher
natural
biological source:
Trigonella foenum-graecum L.
food allergen:
no known allergens
Recommended Products
biological source
Trigonella foenum-graecum L.
Quality Level
grade
FG
Halal
Kosher
natural
reg. compliance
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20
refractive index
n20/D 1.491 (lit.)
density
0.982 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
herbaceous; sweet
Preparation Note
Extraction method: solvent extraction
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
165.2 °F - closed cup
Flash Point(C)
74 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service