Key Documents
SYNF301925
Synthware single-neck round-bottom flask
25 mL, joint: ST/NS 19/22
Synonym(s):
Synthware F301925, glass round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F301925
capacity
25 mL
25 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.