Skip to Content
Merck
All Photos(4)

Documents

N9780

Sigma-Aldrich

2-Nitroaniline

98%

Synonym(s):

1-Amino-2-nitrobenzene, 1-Nitro-2-aminobenzene, 2-Aminonitrobenzene, 2-Nitrobenzenamine, 2-Nitrophenylamine, o-Aminonitrobenzene, o-Nitroaniline, o-Nitrobenzenamine, o-Nitrophenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NH2
CAS Number:
Molecular Weight:
138.12
Beilstein:
509275
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

bp

284 °C (lit.)

mp

70-73 °C (lit.)

SMILES string

Nc1ccccc1[N+]([O-])=O

InChI

1S/C6H6N2O2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H,7H2

InChI key

DPJCXCZTLWNFOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

332.6 °F - closed cup

Flash Point(C)

167.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulia S Sotnikova et al.
Talanta, 220, 121400-121400 (2020-09-16)
The properties of chromatographic columns are largely determined by functional groups located on the sorbent surface. For monolithic columns, surface functional groups can be created during synthesis stage or by chemical bonding with the complete surface of the sorbent. One
Weiwei Qin et al.
Journal of chromatography. A, 1432, 84-91 (2016-01-21)
Metal-organic framework (MOF) MIL-100(Fe) with well-defined thickness was homogenously coated onto the outer surface of magnetic microparticles via a liquid-phase epitaxy method. The as-synthesized MIL-100(Fe) was used as stationary phase for high-performance liquid chromatography (HPLC) and separations of two groups
Min-Quan Yang et al.
Nanoscale, 7(43), 18062-18070 (2015-09-26)
Considerable effort has been made to fabricate graphene (GR)/semiconductor composite photocatalysts, by using graphene oxide (GO) as the most widely used precursor of GR, toward an improved efficacy of solar energy conversion. However, thus far, the role of GO in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service