GF39959364
Polystyrene - Conductive
(high impact conductive polystyrene), sheet, thickness 3 mm, size 150 × 150 mm, black
Synonym(s):
ST323300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
sheet
manufacturer/tradename
Goodfellow 399-593-64
size
150 × 150 mm
thickness
3 mm
color
black
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service