DWKWCL0350-1
WHEATON® CELLine Multiuse Bioreactor flasks for suspension cell culture
polystyrene, flask capacity (350 mL), case of 1 ea
All Photos(1)
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
Recommended Products
material
molded polypropylene neck
polystyrene
screw top plastic cap
sterility
sterile; γ-irradiated
feature
with cap
packaging
case of 1 ea
manufacturer/tradename
WHEATON® WCL0350-1
flask capacity
(350 mL)
Related Categories
General description
CELLine Bioreactor Flask for high density suspension cell culture.
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines
- Disposable and ready-to-use
- High call density and high product concentration
- Bulk media chamber allows for longer run times without media refreshing and reduces operation time
- Decreases use of consumables
- Cost-efficient, space saving, and stackable
- No additional equipment required for operation
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)