DWK213902402
DURAN® measuring cylinder with blue scale ring graduations and hexagonal base
glass, cylinder capacity (100 mL), class A, with certificate
Synonym(s):
213902402
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
glass
feature
class A with certificate
packaging
case of 2 ea
manufacturer/tradename
DURAN® 213902402
cylinder capacity
(100 mL)
Looking for similar products? Visit Product Comparison Guide
General description
DURAN® Measuring cylinders are for holding measuring varying amounts of liquids. Available in two different accuracy Classes – Class A with main ring points graduation and Class B with line graduation. For additional differentiation, the measuring cylinders with accuracy Class A have a blue graduation whereas the B Class measuring cylinders a white graduation. Volume content tolerances for measuring and mixing cylinders conform to DIN and ISO accuracy limits. Thus the desired concentration can be precisely created. Measuring cylinders with a batch number and accuracy Class A are supplied with a batch certificate. This certificate documents the mean value obtained from measuring the batch in question, the standard deviation and the day of issue. The batch certificates can also be retrieved online. The batch number consists of four digits. The first two numbers specify the production year, and the following two numbers specify the batch, e.g.: 18.01.
Features and Benefits
• Complies with ISO 4788• Manufactured from borosilicate glass 3.3• Large hexagonal base equipped with three knobs which increase its stability and prevent the cylinder from rolling• A uniform wall thickness over the entire measuring range prevents wedge errors• Calibration is based on the poured in volume (“In”) at a + 20 °C reference temperature• The volume content tolerances conform to accuracy Class A• Easy-to-read scale with graduation marks to estimate the content• With circular ring graduations• Fired-on, highly durable blue print
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service