Skip to Content
Merck
All Photos(1)

Documents

C93800

Sigma-Aldrich

3-Cyanophenol

99%

Synonym(s):

3-Hydroxybenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4OH
CAS Number:
Molecular Weight:
119.12
Beilstein:
2041515
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

78-81 °C (lit.)

SMILES string

Oc1cccc(c1)C#N

InChI

1S/C7H5NO/c8-5-6-2-1-3-7(9)4-6/h1-4,9H

InChI key

SGHBRHKBCLLVCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service