Documents
BR6729
Discharge valve for Dispensette® S
Organic, 5 and 10 mL, (PFA/Boro 3.3/ceramic/tantalum), valve marking with "ORG"
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
(PFA/Boro 3.3/ceramic/tantalum)
manufacturer/tradename
BRAND 6729
packaging
1 ea of
capacity
5 and 10 mL
General description
Discharge Valve
Legal Information
Dispensette is a registered trademark of BRAND GMBH + CO KG