Key Documents
BR51422
BRAND® ground joint sleeve
PTFE, joint: 29/32, thin-walled
Synonym(s):
PTFE
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
material
PTFE
Joint
joint: 29/32
feature
thin-walled
manufacturer/tradename
BRAND 51422
General description
Made from PTFE, this greaseless seal ensures that samples remain uncontaminated. It offers exceptional chemical resistance across a wide temperature range, from -200 to +260 °C. The thin-walled sleeves, measuring just 0.05 mm thick, fit snugly without adding bulk. The joints are designed for a perfect fit without a protruding rim, simplifying the use of joint clips. This versatile product is suitable for a wide range of applications, including vacuum conditions down to 0.1 mbar, making it ideal for use with rotary evaporators.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2