Key Documents
BR25900
BRAND® pipette controller micro-classic
Synonym(s):
Pipette Controller, Pipetting Aids, micro Pipette Controller
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
(Plastic & metal with autoclavable tube)
packaging
pack of 1 ea
manufacturer/tradename
BRAND 25900
volume
1 mL
General description
Working under a microscope requires utmost concentration and therefore comfortable and reliable instruments. The micro-classic pipette controller with its ergonomic shape and simple handling offers comfort and convenience for this strenuous job. It is a must in IVF and medical laboratories. Suitable for disposable micropipettes with ring mark and other small volume pipettes up to 1 mL (e.g., blood diluting pipettes). The micro-classic adapts to right- and left-handed operation. Adapter and suction tube are autoclavable at 121 °C. The micro-classic minimizes the risk of contamination when working with infectious material. includes 2 spare suction tubes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)