Skip to Content
Merck
All Photos(1)

Key Documents

939544

Sigma-Aldrich

(S)-tBuPHOX-Pd-MAH

new

Synonym(s):

(S)-tert-Butyl-2-[2-(diphenylphosphino)phenyl]-4,5-dihydrooxazole-palldium-maleicanhydride, (S)-tBuPHOX-Pd-maleic-anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H28NO4PPd
CAS Number:
Molecular Weight:
591.93
UNSPSC Code:
12352100

form

powder

Quality Level

reaction suitability

core: palladium
reaction type: Trost Asymmetric Allylic Alkylation
reaction type: Tsuji-Trost Reaction
reagent type: catalyst

color

faint beige to dark beige-brown

SMILES string

CC(C)(C)[C@@H]1[N]([Pd]23C4([H])=C3([H])C(OC4=O)=O)=C(C5=CC=CC=C5[P]2(C6=CC=CC=C6)C7=CC=CC=C7)OC1

InChI

InChI=1S/C25H26NOP.C4H2O3.Pd/c1-25(2,3)23-18-27-24(26-23)21-16-10-11-17-22(21)28(19-12-6-4-7-13-19)20-14-8-5-9-15-20;5-3-1-2-4(6)7-3;/h4-17,23H,18H2,1-3H3;1-2H;/q;;-1/p+1/t23-;;/m1../s1

InChI key

FGSJOEKZZULTLF-MQWQBNKOSA-O

General description

(S)-tBuPHOX-Pd-MAH is a chiral Pd(0) precatalyst first produced in the group of David C. Leitch for asymmetric allylic alkylation reactions. The complex bears a chiral Pfaltz-Helmchen-Williams (PHOX) ligand as well as an easily diplaced maleic anhydride (MAH) ligand.

Features and Benefits

(S)-tBuPHOX-Pd-MAH is a rare example of a chiral, isolable Pd(0) precatalyst for allylic alkylation reactions. Using a chiral single-component precatalyst instead of forming the chiral catalytic speices in-situ avoids activity, reproducibility, and/or selectivity problems arising from incomplete formation of the desired chiral catalyst.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingjun Huang et al.
Nature communications, 14(1), 8058-8058 (2023-12-06)
Stereoselective carbon-carbon bond formation via palladium-catalyzed asymmetric allylic alkylation is a crucial strategy to access chiral natural products and active pharmaceutical ingredients. However, catalysts based on the privileged Trost and Pfaltz-Helmchen-Williams PHOX ligands often require high loadings, specific preactivation protocols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service