Skip to Content
Merck
All Photos(1)

Key Documents

926345

Sigma-Aldrich

NanoFabTx-DC-Chol Lipid Mix

for synthesis of cationic (DC-cholesterol) liposomes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

storage temp.

−20°C

General description

NanoFabTx-DC-Chol Lipid Mix; for synthesis of cationic (DC-cholesterol) liposomes is a ready-to-use nanoformulation blend for the synthesis of liposomes for drug delivery. The NanoFabTx-DC-Chol Lipid Mix includes optimized protocols with step-by-step instructions for synthesizing cationic DC-cholesterol liposomes for drug delivery applications. The modification of the liposomes with the cationic lipid, DC-cholesterol, have numerous advantages including high gene transfection efficiency. Liposome-based formulations are widely used for drug delivery applications and enable improved therapeutic efficacy of a range of drug types including small molecules, nucleic acids, proteins, and peptides.

Application

About NanoFabTx

NanoFabTx lipid mixes and formulation kits enable users to encapsulate a wide variety of therapuetic drug molecules for targeted or extended drug delivery without the need for lengthy trial-and-error optimization. NanoFabTx reagent kits provide an easy-to-use toolkit for encapsulating a variety of therapeutics in nanoparticles, microparticles, or liposomes. Drug encapsulated particles synthesized with the NanoFabTx kits are suitable for biomedical research applications such as oncology, immuno-oncology, gene delivery, and vaccine delivery.

Features and Benefits

  • A ready-to-use nanoformulation blend for the synthesis of cationic DC-cholesterol liposomes
  • Step-by-step protocols (extrusion or microfluidic) developed and tested by our formulation scientists
  • Flexible synthesis tools to create uniform and reproducible liposomes
  • Optimized to make liposomes around 100 nm with low polydispersity
  • DC-Cholesterol allows for high transfection efficiency and targeted drug delivery
Comprehensive protocols for liposome synthesis are included:
  • A lipid film hydration and extrusion protocol.
  • A microfluidics protocol using commercial platforms or syringe pumps.
The microfluidics protocol included with this product uses the NanoFabTx device kit (911593). These kits are ready-to-use and include the microfluidic chip, fittings, and tubing required for microfluidics-based synthesis (compatible microfluidics system or syringe pump required).

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances on liposomal nanoparticles: synthesis, characterization and biomedical applications
Panahi Y, et al.
Artificial Cells, Nanomedicine, and Biotechnology (Print), 45, 788-799 (2017)
Sterically stabilized liposomes: improvements in pharmacokinetics and antitumor therapeutic efficacy
Papahadjopoulos D, et al.
Proceedings of the National Academy of Sciences of the USA, 88, 11460-11464 (1991)
Characterization of liposomal systems containing doxorubicin entrapped in response to pH gradients.
Mayer LD, et al.
Biochimica et Biophysica Acta, 1025, 143-151 (1990)
Which polymers can make nanoparticulate drug carriers long-circulating?
Torchilin VP, et al.
Advanced Drug Delivery Reviews, 16, 141?155-141?155 (1995)
A L Klibanov et al.
FEBS letters, 268(1), 235-237 (1990-07-30)
Incorporation of dioleoyl N-(monomethoxy polyethyleneglycol succinyl)phosphatidylethanolamine (PEG-PE) into large unilamellar liposomes composed of egg phosphatidylcholine:cholesterol (1:1) does not significantly increase the content leakage when the liposomes are exposed to 90% human serum at 37 degrees C, yet the liposomes show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service