Skip to Content
Merck
All Photos(5)

Key Documents

920967

Sigma-Aldrich

Monolayer hexagonal Boron Nitride (hBN) on copper foil

size 3 in. (3 in.) × 76 mm (76 mm)

Synonym(s):

Boronitrene, Hexagonal boron nitride monolayer, Single-layer hexagonal boron nitride, White graphene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
h-BN
CAS Number:
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.23

description

Bandgap: 5.97 eV
Monolayer h-BN
Raman Peak: 1370 /cm-1
h-BN Coverage: 100% with sporadic adlayers

Quality Level

size

3 in. (3 in.) × 76 mm (76 mm)

grain size

>4 μm

SMILES string

B#N

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For all hBN-on-copper products, the displayed range represents electronic data that were obtained after transfer hBN to SiO2. Your own metrics will depend entirely on the transfer methods that you use, and the resultant quality of your transfers.

Application

Monolayer hexagonal boron nitride (h-BN), also known as “white graphene”, is a wide-bandgap 2D crystal (~6 eV that can be tuned to ~2 eV) with exceptional strength , large oxidation resistance at high temperatures , and optical functionalities.
Among its potential applications are:
  • Two-dimensional electronics
  • Nanophotonic and other optoelectronic devices †††
  • Quantum communication and information science
  • Aerospace industry
  • MEMS and NEMS
  • Micro-/nano- actuators
  • Insulating/transparent coatings

Preparation Note

Which side contains hBN? Both sides of the copper foil contain hBN. We primarily recommend that the customer only use the material on the top side (i.e. the side that has Kapton tape) because it is the only side that is guaranteed not to have been in physical contact with anything and therefore should not have any potential physical damage. However, the underside can be used if one is careful and doesn’t mind the damage potential.

Storage and Stability

To ensure the maximum shelf life of your hBN sample, it is best stored under vacuum or in inert atmosphere (Argon or Nitrogen) conditions once the vacuum sealed package has been opened.

Substrates

3” x 3” h-BN/copper foil pieces are secured to their underlying plastic containers with four small Kapton tape pieces, one in each corner. Gently peel the tape or cut off the corners of the foil to release the foil from the plastic container.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hexagonal boron nitride nanomechanical resonators with spatially visualized motion
Zheng Xu, et al.
Microsystems & Nanoengineering, 3 (2017)
Chemical and Bandgap Engineering in Monolayer Hexagonal Boron Nitride
Ba K, et al.
Scientific reports, 7 (2017)
From 2-D to 0-D Boron Nitride Materials, The Next Challenge
Stagi L, et al.
Materials, 12(23) (2019)
Piezoelectricity in Monolayer Hexagonal Boron Nitride
Ares P, et al.
Advanced Materials, 32 (2020)
Monolayer to Bulk Properties of Hexagonal Boron Nitride
Wickramaratne D, et al.
Physical Chemistry, 122, 25524-25529 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service