Skip to Content
Merck
All Photos(1)

Documents

920053

Sigma-Aldrich

Graphene, monolayer film

large grain, CVD graphene on copper foil, A3 size

Synonym(s):

Roll-to-Roll graphene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
CAS Number:
UNSPSC Code:
12141908
NACRES:
NA.23

product name

R2R Monolayer large grain CVD graphene on copper foil, A3 size, avg. no. of layers, 1

Quality Level

description

Growth method: roll-to-roll CVD
Number of layer: Monolayer
Raman intensity 2D/G: ≥1.5

feature

avg. no. of layers 1

sheet resistance

280 Ω/sq ±10%

size

110 μm × 110 μm ± 10% , grain size

thickness

35 μm , Cu Foil

surface coverage

surface coverage >98%

transmittance

>97%

semiconductor properties

(mobility>3000 cm2/V·s) (Hall effect measurements)

Looking for similar products? Visit Product Comparison Guide

General description

Roll-to-roll, high-quality, monolayer CVD graphene with large grain size (∼110μm2) on copper foil, A3 size.

Application

Our Roll-to-Roll CVD graphene products are true monolayer high quality graphene, fabricated inside a Cleanroom, heavily monitored and QC to assure high reproducibility.
The roll-to-roll process allows continuous, large scale graphene production.

This large grain size product with low sheet resistance would enable unmatched reproducibility and allow high performance for CVD graphene based FET, CVD graphene based sensors, and heterostructure based micro/nano electronics.

Application examples:
  • Ultrafast Transistor
  • Optical devices
  • Bio/Gas Sensor
  • Transparent Electrode
  • Flexible Display
  • Smart Coating
  • Thermal management

Storage and Stability

Avoid direct sun light, avoid high temperature, avoid high humidity, and avoid dust.
Be cautious not to drop
Keep away from contamination, heat, dust and flame etc.

Legal Information

Product of LG Electronics, R&D use only

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao-Ling Tang et al.
ACS nano, 11(12), 12817-12823 (2017-11-29)
Two-dimensional (2D) materials are drawing growing attention for next-generation electronics and optoelectronics owing to its atomic thickness and unique physical properties. One of the challenges posed by 2D materials is the large source/drain (S/D) series resistance due to their thinness
Bing Deng et al.
Advanced materials (Deerfield Beach, Fla.), 31(9), e1800996-e1800996 (2018-10-03)
Chemical vapor deposition (CVD) is considered to be an efficient method for fabricating large-area and high-quality graphene films due to its excellent controllability and scalability. Great efforts have been made to control the growth of graphene to achieve large domain
Integrating graphene into semiconductor fabrication lines.
Daniel Neumaier et al.
Nature materials, 18(6), 525-529 (2019-05-23)

Articles

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service