Skip to Content
Merck
All Photos(1)

Key Documents

916404

Sigma-Aldrich

Silver nitrate

99.995% trace metals basis

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Assay:
99.995% trace metals basis
form:
solid

vapor density

5.8 (vs air)

Quality Level

Assay

99.995% trace metals basis

form

solid

mp

212 °C (dec.) (lit.)

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nitrate is a white crystalline salt, and it is less light-sensitive than other silver halides.. It can simply be produced by dissolving silver in nitric acid and then evaporating the solution.

Application

Silver nitrate can be used:
  • As a lithium storage material for lithium-ion batteries due to its lithiation/delithiation behavior.
  • To fabricate Ag decorated rGO electrocatalyst for the oxygen reduction reaction in zinc air batteries.
  • As an electron sacrificial reagent for overall water splitting reaction.
  • As a precursor to tune the shape of gold nanoparticles.
  • To prepare silver vanadium oxide cathode material for batteries.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silver vanadium oxides and related battery applications
Kenneth J. Takeuchi
Coordination Chemistry Reviews, 219-221, 283-310 (2001)
Lithiation/delithiation behavior of silver nitrate as lithium storage material for lithium-ion batteries
Peng Li, et al.
ACS sustainable chemistry & engineering, 5, 5686-5693 (2017)
Silver decorated reduced graphene oxide as electrocatalyst for zinc--air batteries
Laksanaporn Poolnapol, et al.
ENERGIES, 13, 462-462 (2020)
Water oxidation of visible-light-responsive bismuth-yttrium oxychloride promoted by a dual-ion doping strategy for assembly of Z-scheme overall water splitting
Mingxu Hu, et al.
Journal of Materials Chemistry, 10, 16541-16546 (2022)
Understanding the role of AgNO3 concentration and seed morphology in the achievement of tunable shape control in gold nanostars
Supriya Atta, et al.
Nanoscale, 11, 2946-2958 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service