914118
Titania nanoparticles
dispersion, 5 wt. % in ethanol, 700 nm particle size
Synonym(s):
TiO2 nanoparticles, TiOxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
dispersion
nanoparticles
Quality Level
concentration
5 wt. % in ethanol
particle size
700 nm
InChI
1S/2O.Ti
InChI key
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Titania nanoparticles are useful for a wide variety of applications, due to their high refractive index and large bandgap, providing optical properties in the visible spectrum. Applications of titania nanoparticles range from energy to medical applications.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
49.5 °F
Flash Point(C)
9.7 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Modified titanium oxide (TiO2) nanocomposites and its array of applications: a review
Toxicological & Environmental Chemistry, 491-514 (2015)
Introduction: titanium dioxide (TiO2) nanomaterials
Chemical Reviews, 114(19), 9281-9282 (2014)
Mini review on photocatalysis of titanium dioxide nanoparticles and their solar applications.
Nano Energy, 2(5), 1031-1045 (2013)
Nanoscale research letters, 12(1), 225-225 (2017-03-30)
Cancer has become a global problem. On all continents, a great number of people are diagnosed with this disease. In spite of the progress in medical care, cancer still ends fatal for a great number of the ill, either as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service