Skip to Content
Merck
All Photos(1)

Key Documents

900731

Sigma-Aldrich

Nitric acid-d solution

98 atom % D, 90 wt. % in D2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DNO3
CAS Number:
Molecular Weight:
64.02
MDL number:
UNSPSC Code:
12352106

isotopic purity

98 atom % D

form

solution

concentration

90 wt. % in D2O

mass shift

M+1

storage temp.

−20°C

SMILES string

[2H]O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)/i/hD

InChI key

GRYLNZFGIOXLOG-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service