Skip to Content
Merck
All Photos(2)

Documents

855456

Sigma-Aldrich

D-Tyrosine

ReagentPlus®, 99%

Synonym(s):

(R)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-D-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
Beilstein:
2212157
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

optical activity

[α]20/D +10.3°, c = 5 in 1 M HCl

optical purity

ee: 99% (GLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

N[C@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m1/s1

InChI key

OUYCCCASQSFEME-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofeng Fang et al.
Developmental cell, 48(3), 371-382 (2019-01-01)
As integral regulators in plant development and stress response, microRNAs (miRNAs) themselves need to be tightly regulated. Here, we show that tocopherols (vitamin E), lipid-soluble antioxidants synthesized from tyrosine in chloroplasts, positively regulate the biogenesis of miRNAs. Tocopherols are required
Alessio Innocenti et al.
Bioorganic & medicinal chemistry letters, 20(5), 1701-1704 (2010-02-05)
The protein encoded by the NCE103 gene of Candida glabrata, a beta-carbonic anhydrase (CA, EC 4.2.1.1) designated as CgCA, was investigated for its activation with amines and amino acids. CgCA was weakly activated by amino acids such as l-/d-His, l-Phe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service