Skip to Content
Merck
All Photos(1)

Documents

80077

Sigma-Aldrich

1-(4-Trifluoromethylphenyl)piperazine

≥98.0% (GC)

Synonym(s):

1-(α,α,α-Trifluoro-p-tolyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13F3N2
CAS Number:
Molecular Weight:
230.23
Beilstein:
523408
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

crystals

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

88-92 °C

SMILES string

FC(F)(F)c1ccc(cc1)N2CCNCC2

InChI

1S/C11H13F3N2/c12-11(13,14)9-1-3-10(4-2-9)16-7-5-15-6-8-16/h1-4,15H,5-8H2

InChI key

IBQMAPSJLHRQPE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service