Skip to Content
Merck
All Photos(1)

Key Documents

798347

Sigma-Aldrich

Exo-2-Naphthyl Kwon [2.2.1] Bicyclic Phosphine

Synonym(s):

(1S,4S,5R)-5-(naphthalen-2-yl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22NO2PS
Molecular Weight:
395.45
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

153-158 °C

functional group

phosphine
sulfonamide

storage temp.

2-8°C

SMILES string

O=S(N1C[C@H]2[P@](C3=CC=C4C(C=CC=C4)=C3)C[C@@H]1C2)(C5=CC=C(C)C=C5)=O

InChI

1S/C22H22NO2PS/c1-16-6-10-22(11-7-16)27(24,25)23-14-21-13-19(23)15-26(21)20-9-8-17-4-2-3-5-18(17)12-20/h2-12,19,21H,13-15H2,1H3

InChI key

SJQXCVUHXMHJMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The bicyclic, chiral phosphine was developed by the Kwon Research Group.Its initial applications were for asymmetric [3+2] annulation between allenes and imines as well as the first examples of phosphine-catalyzed asymmetric syntheses of 1,2,3,5-substituted pyrrolines. Along with nucleophilic organocatalysis, the P-chiral phosphines may also find utility in asymmetric transition-metal catalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiting Gu et al.
Journal of the American Chemical Society, 137(19), 6400-6406 (2015-04-24)
Two classes of phosphine-catalyzed addition/cycloaddition domino reactions of β'-acetoxy allenoate 1 have been developed. The reaction of 1 with 2-acyl-3-methyl-acrylonitrile 2 readily occurs to give 2-oxabicyclo[3.3.1]nonane 3, furnishing the β'-addition/[4 + 4] cycloaddition domino sequence. In this sequence, β'C of
Ian P Andrews et al.
Chemical science, 3(8), 2510-2514 (2012-07-17)
In this study we performed the total synthesis of the terpene indole alkaloid (+)-ibophyllidine through a pathway involving asymmetric phosphine catalysis, with our novel l-4-hydroxyproline-derived chiral phosphine mediating the key [3 + 2] annulation. Hydrogenation of the [3 + 2]
Christopher E Henry et al.
Journal of the American Chemical Society, 136(34), 11890-11893 (2014-08-08)
We have prepared two new diastereoisomeric 2-aza-5-phosphabicyclo[2.2.1]heptanes from naturally occurring trans-4-hydroxy-L-proline in six chemical operations. These syntheses are concise and highly efficient, with straightforward purification. When we used these chiral phosphines as catalysts for reactions of γ-substituted allenoates with imines

Articles

Chiral phosphines have been the staple ligands for asymmetric transition metal catalysis and more recently operate as catalysts in organic phosphinocatalysis.

Related Content

The Kwon Group has made major strides in the development and application of nucleophilic phosphinocatalysis reactions as a means to synthesize carbo- and heterocycles that serve as synthetic intermediates for both natural products and medicinally useful compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service