Skip to Content
Merck
All Photos(1)

Documents

712221

Sigma-Aldrich

Boc-propargyl-Gly-OH

≥98.0% (HPLC)

Synonym(s):

(S)-2-(Boc-amino)-4-pentynoic acid, Boc-L-2-propargylglycine, Boc-Pra-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO4
CAS Number:
Molecular Weight:
213.23
Beilstein:
5739897
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

optical activity

[α]/D 26.0±2.0°, c = 1 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis
reaction type: click chemistry

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CC#C)C(O)=O

InChI

1S/C10H15NO4/c1-5-6-7(8(12)13)11-9(14)15-10(2,3)4/h1,7H,6H2,2-4H3,(H,11,14)(H,12,13)/t7-/m0/s1

InChI key

AMKHAJIFPHJYMH-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service