Skip to Content
Merck
All Photos(1)

Key Documents

660655

Sigma-Aldrich

D-Glucose-1-13C

endotoxin tested, 99 atom % 13C

Synonym(s):

Labeled Glucose, Dextrose-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
Beilstein:
2263682
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

suitability

endotoxin tested

mass shift

M+1

SMILES string

OC[C@H]1O[13CH](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i6+1

InChI key

WQZGKKKJIJFFOK-USBRANDWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug,household, or other uses. Any other use is the sole responsibility of the enduser.Endotoxin testing is done on the bulk form of the product before subdivisionpackaging. This bulk test does not guarantee that the subdivision or repackagedaliquot is endotoxin-free when it is received or used by the customer and doesnot imply suitability for any particular purpose. If the product must beendotoxin-free for the intended application, the product should be tested priorto actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingjye Jiang et al.
Nucleic acids research, 39(20), 8992-9008 (2011-07-21)
Iron-inducible transcription of the ap65-1 gene in Trichomonas vaginalis involves at least three Myb-like transcriptional factors (tvMyb1, tvMyb2 and tvMyb3) that differentially bind to two closely spaced promoter sites, MRE-1/MRE-2r and MRE-2f. Here, we defined a fragment of tvMyb2 comprising

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service