Skip to Content
Merck
All Photos(3)

Documents

633631

Sigma-Aldrich

Cobalt aluminum oxide

nanopowder, <50 nm particle size (BET), ≥99% trace metals basis (BET)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al2CoO4
CAS Number:
Molecular Weight:
176.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99% trace metals basis (BET)

form

nanopowder

particle size

<50 nm (BET)

SMILES string

[Co++].[O-][Al]=O.[O-][Al]=O

InChI

1S/2Al.Co.4O/q;;+2;;;2*-1

InChI key

BVTIIQXILLBFIK-UHFFFAOYSA-N

Legal Information

Product of Engi-Mat Co.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

So Young Kim et al.
Toxicology in vitro : an international journal published in association with BIBRA, 65, 104813-104813 (2020-03-03)
Phototoxicity due to dermally impregnated compounds exposed to ultraviolet radiation is associated with skin inflammation. The phototoxicity potential of active substances applied to the skin should be evaluated. Pigments are widely used for tattoos, and hypersensitivity reactions, such as photoallergic

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service