Skip to Content
Merck
All Photos(1)

Key Documents

55088

Sigma-Aldrich

8-Hydroxy-2-quinolinecarboxylic acid

≥98.0% (HPLC)

Synonym(s):

8-Hydroxyquinaldic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO3
CAS Number:
Molecular Weight:
189.17
Beilstein:
146082
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

solid

functional group

carboxylic acid

SMILES string

OC(=O)c1ccc2cccc(O)c2n1

InChI

1S/C10H7NO3/c12-8-3-1-2-6-4-5-7(10(13)14)11-9(6)8/h1-5,12H,(H,13,14)

InChI key

UHBIKXOBLZWFKM-UHFFFAOYSA-N

General description

8-Hydroxy-2-quinolinecarboxylic acid (8HQC) is a tridentate chelating agent. It reacts with 2-aminophenol to form a benzoxazole derivative, which can undergo fluorescence quenching in water, making it a viable candidate for developing a probe for detecting water in aprotic organic solvents. The analysis of mid-IR and Raman spectra of 8HQC shows the presence of seven tautomers.

Application

8-Hydroxy-2-quinolinecarboxylic acid may be used in the preparation of bis (8-hydroxyquinoline-2-carboxylato-Κ3N,O,O′) cobalt (II) trihydrate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Katarzyna Walczak et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-09)
8-Hydroxyquinaldic acid, the end-metabolite of tryptophan, is well-known metal chelator; however, its role in humans, especially in cancer promotion and progression, has not been fully revealed. Importantly, 8-hydroxyquinaldic acid is the analog of kynurenic acid with evidenced antiproliferative activity towards
Determination of water content in aprotic organic solvents using 8-hydroxyquinoline based fluorescent probe.
Kim JS, et al.
Bull. Korean Chem. Soc., 27(12), 2058-2058 (2006)
Bis (8-hydroxyquinoline-2-carboxylato-?3N,O,O') cobalt (II) trihydrate.
Okabe N and Muranishi Y.
Acta Crystallographica Section E, Structure Reports Online, 58(7), m352-m353 (2002)
A combined experimental and theoretical study on 8-hydroxy-2-quinolinecarboxylic acid.
Badoglu S and Yurdakul S.
Optics and Spectroscopy, 116(2), 196-206 (2014)
Francesco Bellia et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(70), 16690-16705 (2020-07-07)
Metal dysregulation, oxidative stress, protein modification, and aggregation are factors strictly interrelated and associated with neurodegenerative pathologies. As such, all of these aspects represent valid targets to counteract neurodegeneration and, therefore, the development of metal-binding compounds with other properties to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service