Skip to Content
Merck
All Photos(1)

Key Documents

543993

Sigma-Aldrich

N-(5-Hexynyl)phthalimide

≥95%

Synonym(s):

6-Phthalimido-1-hexyne, N-Hex-5-ynylphthalimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NO2
CAS Number:
Molecular Weight:
227.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥95%

Quality Level

Assay

≥95%

mp

68-72 °C (lit.)

functional group

imide

SMILES string

O=C1N(CCCCC#C)C(=O)c2ccccc12

InChI

1S/C14H13NO2/c1-2-3-4-7-10-15-13(16)11-8-5-6-9-12(11)14(15)17/h1,5-6,8-9H,3-4,7,10H2

InChI key

WALVSJIWVWDFCU-UHFFFAOYSA-N

General description

N-(5-Hexynyl)phthalimide is an N-alkyl substituted phthalimide derivative.

Application

N-(5-Hexynyl)phthalimide may be used as a precursor to form silicon-carbon monolayers on silicon nanowires (SiNWs). It may also be used in the preparation of N-(5-(phenylamino)hexyl)phthalimide.
Reactant for:
  • TBAF-catalyzed monoreduction
  • Ruthenium-catalyzed hydrative conjugative addition to alkenes
  • Regioselective nickel-catalyzed carbocyanation reactions
  • Zinc-catalyzed intermolecular hydroaminations
  • Copper catalyzed cycloaddition with azides

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General Zinc?Catalyzed Intermolecular Hydroamination of Terminal Alkynes.
Alex K, et al.
ChemSusChem, 1(4), 333-338 (2008)
Selective Biofunctionalization all-(111) Surface Silicon Nanowires
Masood M, et al.
MRS Proceedings, 1236 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service