Skip to Content
Merck
All Photos(1)

Key Documents

521671

Sigma-Aldrich

2,6-Difluorobenzylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2ZnBr
CAS Number:
Molecular Weight:
272.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

0.981 g/mL at 25 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

Fc1cccc(F)c1C[Zn]Br

InChI

1S/C7H5F2.BrH.Zn/c1-5-6(8)3-2-4-7(5)9;;/h2-4H,1H2;1H;/q;;+1/p-1

InChI key

RTRBMULKDJEUPO-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service