All Photos(1)
About This Item
Linear Formula:
Na15NO3
CAS Number:
Molecular Weight:
85.99
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
60 atom % 15N
form
solid
mol wt
85.59 by atom % calculation (60 atom % 15N)
mp
306 °C (lit.)
application(s)
agriculture
mass shift
M+1
SMILES string
[Na+].[O-][15N+]([O-])=O
InChI
1S/NO3.Na/c2-1(3)4;/q-1;+1/i1+1;
InChI key
VWDWKYIASSYTQR-YTBWXGASSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service