Skip to Content
Merck
All Photos(2)

Key Documents

488054

Sigma-Aldrich

Ammonium nitrate-15N2

10 atom % 15N

Synonym(s):

Ammonium-15N nitrate-15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
15NH415NO3
CAS Number:
Molecular Weight:
82.03
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

10 atom % 15N

Quality Level

mol wt

80.24 by atom % calculation (10 atom % 15N)

bp

210 °C (lit.)

mp

169 °C (lit.)

application(s)

agriculture

mass shift

M+2

SMILES string

[15NH3].O[15N+]([O-])=O

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3/i2*1+1

InChI key

PRORZGWHZXZQMV-NPMJFZSBSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service