Skip to Content
Merck
All Photos(1)

Key Documents

480355

Sigma-Aldrich

Poly(dimethylsiloxane), monohydroxy terminated

average Mn ~4,670

Synonym(s):

Terminated PDMS Polymer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~4,670

Quality Level

refractive index

n20/D 1.408 (lit.)

viscosity

~79 cSt(25 °C)(lit.)

density

0.97 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(O)O.CCCC[Si](C)(C)O.C[Si](C)(O)CCCOCCO

InChI

1S/C7H18O3Si.C6H16OSi.C2H8O2Si/c1-11(2,9)7-3-5-10-6-4-8;1-4-5-6-8(2,3)7;1-5(2,3)4/h8-9H,3-7H2,1-2H3;7H,4-6H2,1-3H3;3-4H,1-2H3

Application

Resin modifier and paint additive.

Features and Benefits

Good lubricating properties.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

See All

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service