Skip to Content
Merck
All Photos(1)

Key Documents

474347

Sigma-Aldrich

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate

contains inhibitor, 97%

Synonym(s):

1H,1H,2H,2H-perfluorooctyl acrylate, TFOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CH2(CF2)5CF3
CAS Number:
Molecular Weight:
418.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

contains

HQ, MEHQ as inhibitor

refractive index

n20/D 1.338 (lit.)

bp

76-80 °C/8 mmHg (lit.)

density

1.554 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C

InChI

1S/C11H7F13O2/c1-2-5(25)26-4-3-6(12,13)7(14,15)8(16,17)9(18,19)10(20,21)11(22,23)24/h2H,1,3-4H2

InChI key

VPKQPPJQTZJZDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1 - STOT RE 2

Target Organs

Liver,Teeth

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sita Bierma-Zeinstra et al.
Best practice & research. Clinical rheumatology, 34(2), 101564-101564 (2020-08-11)
Osteoarthritis (OA) is a chronic, disabling, and common disease. Nonpharmacological and nonsurgical treatments are seen as the core treatments for OA. This paper presents an overview of the recommendations from international guidelines on nonpharmacological and nonsurgical interventions. Education, advice, or
Hirotaka Iijima et al.
BMC musculoskeletal disorders, 18(1), 126-126 (2017-03-28)
This 1-year prospective cohort study aimed to compare the changes in clinical symptoms and functional disability between patients with coexisting patellofemoral (PF) and tibiofemoral (TF) osteoarthritis (OA) and those with isolated TFOA. Seventy-two patients with medial knee OA were enrolled.
Yuchen Fu et al.
ACS applied materials & interfaces, 9(35), 30161-30170 (2017-08-15)
The superhydrophobic antibacterial fabrics with intelligent switchable wettability were fabricated by the cross-link reaction among pH-responsive antibacterial copolymer tethered hydroxyl groups, methylol-contained poly(ureaformaldehyde) nanoparticles (PUF NPs), and hexamethylene diisocyanate. It was found that the surface concentration of N
Elena Masotti et al.
Polymers, 12(2) (2020-02-14)
Fluorinated (F6) and zwitterionic, as well as phosphorylcholine (MPC) and sulfobetaine (MSA), copolymers containing a low amount (1 and 5 mol%) of 3-(trimethoxysilyl)propyl methacrylate (PTMSi) were prepared and covalently grafted to glass slides by using the trimethoxysilyl groups as anchorage
Elisa Martinelli et al.
Biofouling, 32(1), 81-93 (2016-01-16)
Block copolymers made from a poly(dimethyl siloxane) (Si) and a poly(meth)acrylate carrying oxyethylene (EG) or fluoroalkyl (AF) side chains were synthesized and incorporated as surface-active components into a silicone matrix to produce cross-linked films with different surface hydrophilicity/phobicity. Near-edge X-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service