Skip to Content
Merck
All Photos(2)

Documents

432776

Sigma-Aldrich

Poly(acrylamide-co-acrylic acid) potassium salt

cross-linked, 200-1000 μm particle size

Synonym(s):

Acrylamide-potassium acrylate copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

crosslinking

cross-linked

composition

potassium, ~9%

particle size

200-1000 μm

pH

5.5-6.0

density

0.54 g/mL at 25 °C (lit.)

SMILES string

NC(=O)C=C.[K]OC(=O)C=C

InChI

1S/C3H5NO.C3H4O2.K/c2*1-2-3(4)5;/h2H,1H2,(H2,4,5);2H,1H2,(H,4,5);/q;;+1/p-1

InChI key

HNXHILBJDFONSX-UHFFFAOYSA-M

Features and Benefits

Superabsorbent polymer. Rapid absorption of water. Material can undergo many wet/dry cycles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service