Skip to Content
Merck
All Photos(1)

Documents

427616

Sigma-Aldrich

Trimethyl borate-11B

99 atom % 11B, 98% (CP)

Synonym(s):

Methyl borate-11B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)311B
CAS Number:
Molecular Weight:
104.11
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 11B

Quality Level

Assay

98% (CP)

refractive index

n20/D 1.358 (lit.)

bp

68-69 °C (lit.)

mp

−34 °C (lit.)

density

0.924 g/mL at 25 °C (lit.)

mass shift

depleted

SMILES string

COB(OC)OC

InChI

1S/C3H9BO3/c1-5-4(6-2)7-3/h1-3H3

InChI key

WRECIMRULFAWHA-UHFFFAOYSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

17.6 °F - closed cup

Flash Point(C)

-8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L V Angelova et al.
Soft matter, 11(25), 5060-5066 (2015-06-02)
Viscoelastic, gel-like, polymeric dispersions (HVPDs) can be prepared by crosslinking polyols with borax or boric acid in water under alkaline conditions. Rheologically similar HVPDs have been prepared in organic liquids containing no water or hydroxylic groups through crosslinking partially or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service