Skip to Content
Merck
All Photos(1)

Key Documents

333557

Sigma-Aldrich

(1-Bromovinyl)trimethylsilane

97%

Synonym(s):

α-(Trimethylsilyl)vinyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(Br)Si(CH3)3
CAS Number:
Molecular Weight:
179.13
Beilstein:
1742258
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

vapor density

>1 (vs air)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.458 (lit.)

bp

124 °C/745 mmHg (lit.)

density

1.156 g/mL at 25 °C (lit.)

functional group

bromo

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C(Br)=C

InChI

1S/C5H11BrSi/c1-5(6)7(2,3)4/h1H2,2-4H3

InChI key

VVDJVCJVVHHCIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrakis(triphenylphosphine)palladium(0) catalyzed coupling reactions of (1-bromovinyl)trimethylsilane with organozinc bromides were studied.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abnormal products of palladium catalysed coupling reactions of (1-bromovinyl) trimethylsilane.
Ennis DS and Gilchrist TL.
Tetrahedron Letters, 30(28), 3735-3736 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service