Skip to Content
Merck
All Photos(1)

Key Documents

329886

Sigma-Aldrich

Water-16O

≥99.94 atom % 16O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H216O
CAS Number:
Molecular Weight:
18.01
EC Number:
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
NACRES:
NA.12

description

17O, 18O-depleted
Normalized with respect to hydrogen

Quality Level

isotopic purity

≥99.94 atom % 16O

form

liquid

pH

6.0-8.0 (25 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 25 °C

mass shift

depleted

SMILES string

[16OH2]

InChI

1S/H2O/h1H2/i1+0

InChI key

XLYOFNOQVPJJNP-IGMARMGPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yareli Rojas-Aguirre et al.
Carbohydrate polymers, 223, 115113-115113 (2019-08-21)
We present three easily rationalized star-shaped PEGylated β-cyclodextrin (βCD) derivatives synthesized via conjugation of different molecular weight PEG chains (5000, 2000, and 550 Da) to the βCD primary face by click chemistry (βCD-PEG5000, βCD-PEG2000, βCD-PEG550 respectively). βCDPEG systems are envisioned to
Emeaga Maheswaran et al.
PloS one, 10(8), e0134967-e0134967 (2015-08-08)
Immunotherapy is emerging as a supplement to conventional cancer treatment, and identifying antigen targets for specific types of cancer is critical to optimizing therapeutic efficacy. Cancer/testis antigens are highly promising targets for immunotherapy due to their cancer-specific expression and antigenic
Masashi Furukawa et al.
PloS one, 10(1), e0118060-e0118060 (2015-01-31)
The lung is one of the most common sites of breast cancer metastasis. While metastatic seeding is often accompanied by a dormancy-promoting mesenchymal to epithelial reverting transitions (MErT), we aimed to determine whether lung epithelial cells can impart this phenotype
Lisa K Lunde et al.
Brain structure & function, 220(4), 2087-2101 (2014-04-30)
Astrocytes are highly polarised cells with processes that ensheath microvessels, cover the brain surface, and abut synapses. The endfoot membrane domains facing microvessels and pia are enriched with aquaporin-4 water channels (AQP4) and other members of the dystrophin associated protein
Liudmila Efremova et al.
British journal of pharmacology, 172(16), 4119-4132 (2015-05-20)
Few neuropharmacological model systems use human neurons. Moreover, available test systems rarely reflect functional roles of co-cultured glial cells. There is no human in vitro counterpart of the widely used 1-methyl-4-phenyl-tetrahydropyridine (MPTP) mouse model of Parkinson's disease We generated such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service